siRNA Details
Gene Symbol: 8430413B15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018406.1 | - | 1 | 204 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720529.1 | - | 1 | 204 | Chr. 6 - 73050646 - 73051173 [+] on Build GRCm38 |
