siRNA Details
Gene Symbol: Gm31333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044531.1 | - | 2, 3 | 200 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB642514.1 | - | 2, 3 | 200 | Chr. 12 - 29141927 - 29198128 [+] on Build GRCm38 |
