siRNA Details
Gene Symbol: Abhd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046757.1 | - | 1 | 2555 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB329581.2 | - | 1 | 175 | Chr. 16 - 45739730 - 45740398 [-] on Build GRCm38 |
