siRNA Details
Gene Symbol: Gm38409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087229.1 | - | 2 | 142 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB664379.1 | - | 2 | 142 | Chr. 14 - 67476873 - 67500553 [+] on Build GRCm38 |
