siRNA Details
Gene Symbol: E030042O20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053216.1 | - | 1 | 692 | ||
AK078602.1 | - | 1 | 596 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV353066.2 | - | 1 | 480 | Chr. 2 - 75704981 - 75705664 [+] on Build GRCm38 |
