siRNA Details
Gene Symbol: 2610209C05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011926.1 | - | 1 | 66 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY711132.1 | - | 1 | 63 | Chr. 6 - 54816779 - 54817648 [-] on Build GRCm38 |
