siRNA Details
Gene Symbol: A430072P03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079802.1 | - | 2 | 417 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB635509.1 | - | 2 | 414 | Chr. 5 - 107726400 - 107728805 [+] on Build GRCm38 |
