siRNA Details
Gene Symbol: Gm16973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046209.1 | 3 | 438 | |||
NR_046210.1 | 3 | 370 | ||||
GenBank mRNA | AK045225.1 | - | 3 | 638 | ||
BC027359.1 | - | 2 | 244 | |||
CO425816.1 | - | 3 | 438 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA466035.1 | - | 4 | 449 | Chr. 14 - 56697772 - 56739394 [-] on Build GRCm38 |
