siRNA Details
Gene Symbol: Gm16973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046209.1 | 3 | 754 | |||
NR_046210.1 | 3 | 686 | ||||
GenBank mRNA | AK045225.1 | - | 3 | 954 | ||
BC027359.1 | - | 2 | 560 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB296680.2 | - | 1 | 300 | Chr. 14 - 56697352 - 56697992 [-] on Build GRCm38 |
