siRNA Details
Gene Symbol: Gm16973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046209.1 | 2, 3 | 400 | |||
NR_046210.1 | 2, 3 | 332 | ||||
GenBank mRNA | AK045225.1 | - | 2, 3 | 600 | ||
BC027359.1 | - | 1, 2 | 206 | |||
CO425816.1 | - | 2, 3 | 400 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA466035.1 | - | 3, 4 | 411 | Chr. 14 - 56697772 - 56739394 [-] on Build GRCm38 |
