siRNA Details
Gene Symbol: 9430034F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK172360.1 | - | 1 | 1137 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU918917.1 | - | 1 | 36 | Chr. 11 - 23713831 - 23714423 [+] on Build GRCm38 |
