siRNA Details
Gene Symbol: Sema3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052051.1 | - | 1 | 129 | ||
AK164242.1 | - | 1 | 27 | |||
AK166127.1 | - | 1 | 206 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sema3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052051.1 | - | 1 | 129 | ||
AK164242.1 | - | 1 | 27 | |||
AK166127.1 | - | 1 | 206 |