siRNA Details
Gene Symbol: 4930570G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040400.1 | 2 | 653 | |||
GenBank mRNA | BC048612.1 | - | 2 | 653 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930570G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040400.1 | 2 | 653 | |||
GenBank mRNA | BC048612.1 | - | 2 | 653 |