siRNA Details
Gene Symbol: 4930570G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040400.1 | 1 | 47 | |||
GenBank mRNA | BC048612.1 | - | 1 | 47 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU961646.1 | - | 50 | Chr. - - [-] on Build |
