siRNA Details
Gene Symbol: 4921519G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014924.1 | - | 1 | 155 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714442.1 | - | 1 | 150 | Chr. 10 - 42855709 - 42856679 [-] on Build GRCm38 |
