siRNA Details
Gene Symbol: 4921520E09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014928.1 | - | 1 | 449 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714444.1 | - | 1 | 445 | Chr. 7 - 30636633 - 30637514 [+] on Build GRCm38 |
