siRNA Details
Gene Symbol: 1700123J03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018982.1 | - | 1 | 186 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707352.1 | - | 1 | 176 | Chr. 12 - 77061026 - 77061247 [-] on Build GRCm38 |
