siRNA Details
Gene Symbol: 1700040B14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006649.1 | - | 1 | 236 | ||
AK018986.1 | - | 1 | 229 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706640.1 | - | 1 | 235 | Chr. 8 - 123518337 - 123518795 [-] on Build GRCm38 |
