siRNA Details
Gene Symbol: E330018M18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087768.1 | - | 2 | 429 | ||
AK141748.1 | - | 2 | 398 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: E330018M18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087768.1 | - | 2 | 429 | ||
AK141748.1 | - | 2 | 398 |