siRNA Details
Gene Symbol: 4833405L11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014656.1 | - | 819 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714152.1 | - | 2 | 819 | Chr. 12 - 29105690 - 29106737 [-] on Build GRCm38 |
