siRNA Details
Gene Symbol: 4831440E17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087767.1 | - | 39 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB666475.1 | - | 1 | 35 | Chr. 5 - 25499070 - 25501165 [+] on Build GRCm38 |
