siRNA Details
Gene Symbol: 9330133O14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045696.1 | 1 | 419 | |||
GenBank mRNA | AK033985.1 | - | 1 | 360 | ||
AK042662.1 | - | 1 | 384 | |||
AK049744.1 | - | 1 | 418 | |||
AK085519.1 | - | 1 | 371 | |||
BC094355.1 | - | 1 | 304 |
