siRNA Details
Gene Symbol: 9330133O14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045696.1 | 1 | 459 | |||
GenBank mRNA | AK033985.1 | - | 1 | 400 | ||
AK042662.1 | - | 1 | 425 | |||
AK049744.1 | - | 1 | 458 | |||
AK085519.1 | - | 1 | 411 | |||
BC094355.1 | - | 1 | 344 |
