siRNA Details
Gene Symbol: Gm9777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019581.1 | - | 2 | 500 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714740.1 | - | 1 | 500 | Chr. 13 - 119408679 - 119409267 [+] on Build GRCm38 |
