siRNA Details
Gene Symbol: 4930428D20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019586.1 | - | 2 | 153 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714774.1 | - | 2 | 153 | Chr. 16 - 69796140 - 69800381 [-] on Build GRCm38 |
