siRNA Details
Gene Symbol: Mgat4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080684.1 | - | 3 | 654 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY294038.1 | - | 3 | 305 | Chr. 10 - 101681836 - 102011938 [+] on Build GRCm38 |
