siRNA Details
Gene Symbol: 4930555G07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016129.1 | - | 2 | 241 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715732.1 | - | 2 | 241 | Chr. 10 - 87230760 - 87239248 [+] on Build GRCm38 |
