siRNA Details
Gene Symbol: 1700091J24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007034.1 | - | 1 | 18 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707102.1 | - | 1 | 18 | Chr. 4 - 133432740 - 133432983 [-] on Build GRCm38 |
