siRNA Details
Gene Symbol: 1700013N06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005966.1 | - | 184 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706098.1 | - | 3 | 184 | Chr. 12 - 106836868 - 106877558 [+] on Build GRCm38 |
