siRNA Details
Gene Symbol: Gm19461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037984.1 | 4 | 3025 | |||
GenBank mRNA | AK145464.1 | - | 5 | 2733 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY756171.1 | - | 2 | 621 | Chr. 1 - 133249852 - 133250598 [-] on Build GRCm38 |
