siRNA Details
Gene Symbol: 9330153N18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020372.1 | - | 1 | 516 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721338.1 | - | 1 | 516 | Chr. 6 - 38610049 - 38610714 [+] on Build GRCm38 |
