siRNA Details
Gene Symbol: 9330154F10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020373.1 | - | 1 | 353 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721342.1 | - | 1 | 350 | Chr. 4 - 68800982 - 68801628 [-] on Build GRCm38 |
