siRNA Details
Gene Symbol: 1700069L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006923.1 | - | 3 | 378 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706925.1 | - | 3 | 378 | Chr. 5 - 113703697 - 113724749 [-] on Build GRCm38 |
