siRNA Details
Gene Symbol: Gm31326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076854.1 | - | 1 | 928 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV265891.2 | - | 1 | 523 | Chr. 9 - 101989662 - 101990307 [+] on Build GRCm38 |
