siRNA Details
Gene Symbol: 1700008K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038141.1 | 2 | 917 | |||
NR_038142.1 | 3 | 820 | ||||
NR_038143.1 | 3 | 826 | ||||
NR_038144.1 | 2 | 773 | ||||
NR_038145.1 | 2 | 656 | ||||
GenBank mRNA | AK005780.1 | - | 3 | 803 |
