siRNA Details
Gene Symbol: 1700008O09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005785.1 | - | 1 | 86 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705939.1 | - | 1 | 86 | Chr. 12 - 106139779 - 106140108 [-] on Build GRCm38 |
