siRNA Details
Gene Symbol: Gm39888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040511.1 | - | 2 | 491 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB590223.2 | - | 2 | 487 | Chr. 2 - 101991980 - 102009016 [+] on Build GRCm38 |
