siRNA Details
Gene Symbol: B230207N12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020997.1 | - | 1 | 532 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI731946.1 | - | 1 | 397 | Chr. 11 - 79703609 - 79704316 [-] on Build GRCm38 |
