siRNA Details
Gene Symbol: Gm29676 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035008.1 | - | 572 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB627242.1 | - | 2 | 572 | Chr. 13 - 44250524 - 44252670 [-] on Build GRCm38 |
