siRNA Details
Gene Symbol: 2900079J23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019356.1 | - | 1 | 64 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712766.1 | - | 1 | 64 | Chr. 13 - 34196617 - 34196884 [-] on Build GRCm38 |
