siRNA Details
Gene Symbol: Scarletltr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040743.1 | 3 | 299 | |||
GenBank mRNA | AK011166.1 | - | 3 | 300 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY710499.1 | - | 3 | 300 | Chr. 9 - 64080761 - 64088046 [+] on Build GRCm38 |
