siRNA Details
Gene Symbol: Gm17597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045688.1 | - | 2 | 307 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE648997.1 | - | 2 | 211 | Chr. 15 - 81801468 - 81802518 [-] on Build GRCm38 |
