siRNA Details
Gene Symbol: 4930431F12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073013.1 | 10 | 1787 | |||
GenBank mRNA | AK015267.1 | - | 10 | 1788 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930431F12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073013.1 | 10 | 1787 | |||
GenBank mRNA | AK015267.1 | - | 10 | 1788 |