siRNA Details
Gene Symbol: 9230104K21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020313.1 | - | 1 | 733 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI696898.1 | - | 1 | 458 | Chr. 4 - 45406256 - 45406914 [-] on Build GRCm38 |
