siRNA Details
Gene Symbol: A730018C14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036459.1 | 1 | 121 | |||
GenBank mRNA | AK042713.1 | - | 1 | 136 | ||
AK048513.1 | - | 1 | 122 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB649310.1 | - | 1 | 120 | Chr. 12 - 112412161 - 112423197 [-] on Build GRCm38 |
