siRNA Details
Gene Symbol: 4930415N12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015151.1 | - | 1 | 318 | ||
AK029619.1 | - | 1 | 318 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV260725.2 | - | 517 | Chr. - - [-] on Build |
