siRNA Details
Gene Symbol: Gm28178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK081439.1 | - | 3 | 235 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB648450.1 | - | 3 | 235 | Chr. 1 - 52572313 - 52580292 [-] on Build GRCm38 |
