siRNA Details
Gene Symbol: Usp29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK083494.1 | - | 1 | 2956 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB442079.2 | - | 1 | 596 | Chr. 7 - 6769493 - 6770139 [+] on Build GRCm38 |
