siRNA Details
Gene Symbol: 2810442N19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040562.1 | 2 | 610 | |||
GenBank mRNA | AK013293.1 | - | 2 | 611 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712295.1 | - | 2 | 611 | Chr. 1 - 162004969 - 162005824 [+] on Build GRCm38 |
