siRNA Details
Gene Symbol: 2210414I22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008926.1 | - | 1 | 226 | ||
AK132110.1 | - | 1 | 355 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ044771.1 | - | 1 | 355 | Chr. 2 - 164911890 - 164912590 [+] on Build GRCm38 |
